missing translation for 'onlineSavingsMsg'
Learn More

7-Octen-1-ol 96.0+%, TCI America™

Supplier:  TCI America O028025ML

Quantity 25 mL

 View more versions of this product

Catalog No. O028025ML


Only null left

Description

Description

Specifications

Chemical Identifiers

13175-44-5
128.215
WXPWPYISTQCNDP-UHFFFAOYSA-N
oct-7-en-1-ol
C8H16O
MFCD00798076
83203
C=CCCCCCCO

Specifications

7-Octen-1-ol
Colorless
C8H16O
MFCD00798076
C=CCCCCCCO
128.215
128.22
Liquid
13175-44-5
66°C
25 mL
WXPWPYISTQCNDP-UHFFFAOYSA-N
oct-7-en-1-ol
83203
≥96.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2393

TSCA : No

SDS
Documents

Documents

Promotions

Promotions