accessibility menu, dialog, popup

UserName

HOCH2

CAS RN 13175-44-5

IUPAC Name: oct-7-en-1-ol
Synonyms: oct-7-en-1-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: WXPWPYISTQCNDP-UHFFFAOYSA-N
SMILES: OCCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results