missing translation for 'onlineSavingsMsg'
Learn More

7-Octen-1-ol 96.0+%, TCI America™

Catalog No. p-7134273
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02805ML 5 mL
O028025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02805ML Supplier TCI America Supplier No. O02805ML

Chemical Identifiers

CAS 13175-44-5
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00798076
InChI Key WXPWPYISTQCNDP-UHFFFAOYSA-N
PubChem CID 83203
IUPAC Name oct-7-en-1-ol
SMILES C=CCCCCCCO

Specifications

Color Colorless
Boiling Point 66°C
Quantity 5 mL
Formula Weight 128.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Octen-1-ol
EINECSNumber (2)-2393
TSCA No