missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1-tetralone 98.0+%, TCI America™

Catalog No. N09455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09455G 5 g
N09451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09455G Supplier TCI America Supplier No. N09455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40353-34-2
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00019661
InChI Key GWAQYWSNCVEJMW-UHFFFAOYSA-N
Synonym 7-nitro-1-tetralone, 7-nitrotetralone, 7-nitro-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-1 2h-naphthalenone, 3,4-dihydro-7-nitronaphthalen-1 2h-one, wln: l66 bvt&j inw, 7-nitro-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone,4-dihydro-7-nitro, 7-nitro-tetralone
PubChem CID 38445
IUPAC Name 7-nitro-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 5 g
Formula Weight 191.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Nitro-1-tetralone
RTECSNumber QK5025000
TSCA No