accessibility menu, dialog, popup

UserName

ONOO

CAS RN 40353-34-2

IUPAC Name: 7-nitro-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 7-nitro-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: GWAQYWSNCVEJMW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2CCCC(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)