missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1-tetralone, 97%

Catalog No. AAL0185304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0185304 2 g
AAL0185309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0185304 Supplier Thermo Scientific Chemicals Supplier No. L0185304
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 40353-34-2 | C10H9NO3 | 191.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40353-34-2
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00019661
InChI Key GWAQYWSNCVEJMW-UHFFFAOYSA-N
Synonym 7-nitro-1-tetralone, 7-nitrotetralone, 7-nitro-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-1 2h-naphthalenone, 3,4-dihydro-7-nitronaphthalen-1 2h-one, wln: l66 bvt&j inw, 7-nitro-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone,4-dihydro-7-nitro, 7-nitro-tetralone
PubChem CID 38445
IUPAC Name 7-nitro-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1

Specifications

Melting Point 103°C to 108°C
Quantity 2 g
Beilstein 1570515
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material 7-Nitro-1-tetralone
EINECSNumber 254-887-6
RTECSNumber QK5025000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only