missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl-8-nitroquinoline, 98+%

Catalog No. p-7043985
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0388206 5 g
AAL0388214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0388206 Supplier Thermo Scientific Chemicals Supplier No. L0388206
Only null left

CAS: 7471-63-8 | C10H8N2O2 | 188.186 g/mol

7-Methyl-8-nitroquinoline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Methyl-8-nitroquinoline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 7471-63-8
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00041234
InChI Key ZZDTVYJYMRSNQL-UHFFFAOYSA-N
Synonym quinoline, 7-methyl-8-nitro, 8-nitro-7-methylquinoline, 7-methyl-8-nitro-quinoline, acmc-1bcox, 7-methyl 8-nitroquinoline
PubChem CID 81981
IUPAC Name 7-methyl-8-nitroquinoline
SMILES CC1=C(C2=C(C=CC=N2)C=C1)[N+](=O)[O-]

Specifications

Melting Point 185°C to 186°C
Quantity 5 g
Beilstein 154601
Solubility Information Insoluble in water.
Formula Weight 188.19
Percent Purity ≥98%
Chemical Name or Material 7-Methyl-8-nitroquinoline
Hazard Category H315-H319-H341
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 231-268-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only