accessibility menu, dialog, popup

UserName

CAS RN 7471-63-8

IUPAC Name: 7-methyl-8-nitroquinoline
Synonyms: 7-methyl-8-nitroquinoline
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: ZZDTVYJYMRSNQL-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=NC2=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)