missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(Diethylamino)coumarin 98.0+%, TCI America™
$100.67 - $329.94
Chemical Identifiers
| CAS | 20571-42-0 |
|---|---|
| Molecular Formula | C13H15NO2 |
| Molecular Weight (g/mol) | 217.27 |
| MDL Number | MFCD00232946 |
| InChI Key | QXAMGWKESXGGNV-UHFFFAOYSA-N |
| PubChem CID | 88598 |
| IUPAC Name | 7-(diethylamino)-2H-chromen-2-one |
| SMILES | CCN(CC)C1=CC=C2C=CC(=O)OC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3354200MG
|
TCI America
D3354200MG |
200 mg |
Each for $100.67
|
|
|||||
|
D33541G
|
TCI America
D33541G |
1 g |
Each for $329.94
|
|
|||||
Chemical Identifiers
| 20571-42-0 | |
| 217.27 | |
| QXAMGWKESXGGNV-UHFFFAOYSA-N | |
| 7-(diethylamino)-2H-chromen-2-one |
| C13H15NO2 | |
| MFCD00232946 | |
| 88598 | |
| CCN(CC)C1=CC=C2C=CC(=O)OC2=C1 |
Specifications
| 20571-42-0 | |
| Yellow | |
| MFCD00232946 | |
| QXAMGWKESXGGNV-UHFFFAOYSA-N | |
| 7-(diethylamino)-2H-chromen-2-one | |
| 88598 | |
| ≥98.0% (GC,T) | |
| 7-(Diethylamino)coumarin |
| 89°C | |
| C13H15NO2 | |
| 200 mg | |
| CCN(CC)C1=CC=C2C=CC(=O)OC2=C1 | |
| 217.27 | |
| 217.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes