accessibility menu, dialog, popup

UserName

H3CNCH3OO

CAS RN 20571-42-0

IUPAC Name: 7-(diethylamino)-2H-chromen-2-one
Synonyms: 7-(diethylamino)chromen-2-one
Molecular Weight (g/mol): 217.27
Molecular Formula: C13H15NO2
InChi Key: QXAMGWKESXGGNV-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=CC(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results