missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(Diethylamino)coumarin 98.0+%, TCI America™
Supplier: TCI America D3354200MG
Specifications
| 7-(Diethylamino)coumarin | |
| 89°C | |
| C13H15NO2 | |
| 200 mg | |
| CCN(CC)C1=CC=C2C=CC(=O)OC2=C1 | |
| 217.27 | |
| 217.27 | |
| Crystalline Powder |
| 20571-42-0 | |
| Yellow | |
| MFCD00232946 | |
| QXAMGWKESXGGNV-UHFFFAOYSA-N | |
| 7-(diethylamino)-2H-chromen-2-one | |
| 88598 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 20571-42-0 | |
| 217.27 | |
| QXAMGWKESXGGNV-UHFFFAOYSA-N | |
| 7-(diethylamino)-2H-chromen-2-one |
| C13H15NO2 | |
| MFCD00232946 | |
| 88598 | |
| CCN(CC)C1=CC=C2C=CC(=O)OC2=C1 |
Safety and Handling
TSCA : Yes