missing translation for 'onlineSavingsMsg'
Learn More

7-Chloropyrazolo[1,5-a]pyrimidine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 58347-49-2
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.57
MDL Number MFCD04035684
InChI Key YCZQHXPIKQHABJ-UHFFFAOYSA-N
Synonym 7-chloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 7-chloro, 7-chloro-pyrazolo 1,5-a pyrimidine, zlchem 1087, 7-chloropyrazolo 1?5-a pyrimidine, 7-chloranylpyrazolo 1,5-a pyrimidine, 7-chloro-8-hydropyrazolo 1,5-a pyrimidine
PubChem CID 12281647
IUPAC Name 7-chloropyrazolo[1,5-a]pyrimidine
SMILES C1=C2N=CC=C(N2N=C1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC438490010
View Documents Promotion Details
N/A 1g Glass bottle N/A
Only null left
 
AC438490050
View Documents Promotion Details
N/A 5g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

58347-49-2
153.57
YCZQHXPIKQHABJ-UHFFFAOYSA-N
12281647
C1=C2N=CC=C(N2N=C1)Cl
C6H4ClN3
MFCD04035684
7-chloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 7-chloro, 7-chloro-pyrazolo 1,5-a pyrimidine, zlchem 1087, 7-chloropyrazolo 1?5-a pyrimidine, 7-chloranylpyrazolo 1,5-a pyrimidine, 7-chloro-8-hydropyrazolo 1,5-a pyrimidine
7-chloropyrazolo[1,5-a]pyrimidine

Specifications

58347-49-2
94% min. (GC)
MFCD04035684
YCZQHXPIKQHABJ-UHFFFAOYSA-N
7-chloropyrazolo[1,5-a]pyrimidine
12281647
95%
7-Chloropyrazolo[1,5-a]pyrimidine
Authentic
C6H4ClN3
7-chloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 7-chloro, 7-chloro-pyrazolo 1,5-a pyrimidine, zlchem 1087, 7-chloropyrazolo 1?5-a pyrimidine, 7-chloranylpyrazolo 1,5-a pyrimidine, 7-chloro-8-hydropyrazolo 1,5-a pyrimidine
C1=C2N=CC=C(N2N=C1)Cl
153.57
153.57
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only