missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1-tetralone 95.0+%, TCI America™

Catalog No. C29761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29761G 1 g
C29765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29761G Supplier TCI America Supplier No. C29761G
Only null left

Chemical Identifiers

CAS 26673-32-5
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.63
MDL Number MFCD01928386
InChI Key IIMAYXKDBHTQHC-UHFFFAOYSA-N
Synonym 7-Chloro-3,4-dihydro-1(2H)-naphthalenone
PubChem CID 256677
IUPAC Name 7-chloro-1,2,3,4-tetrahydronaphthalen-1-one
SMILES ClC1=CC=C2CCCC(=O)C2=C1

Specifications

Melting Point 95°C
Color Yellow
Boiling Point 115°C
Quantity 1 g
Formula Weight 180.63
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloro-1-tetralone