accessibility menu, dialog, popup

UserName

CAS RN 26673-32-5

ClO

CAS RN 26673-32-5

IUPAC Name: 7-chloro-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 7-chloro-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 180.63
Molecular Formula: C10H9ClO
InChi Key: IIMAYXKDBHTQHC-UHFFFAOYSA-N
SMILES: ClC1=CC=C2CCCC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results