missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(Bromomethyl)pentadecane 95.0+%, TCI America™
$72.70 - $212.51
Chemical Identifiers
| CAS | 52997-43-0 |
|---|---|
| Molecular Formula | C16H33Br |
| Molecular Weight (g/mol) | 305.344 |
| MDL Number | MFCD00136231 |
| InChI Key | RWEKWRQKAHQYNE-UHFFFAOYSA-N |
| PubChem CID | 18459580 |
| IUPAC Name | 7-(bromomethyl)pentadecane |
| SMILES | CCCCCCCCC(CCCCCC)CBr |
Chemical Identifiers
| 52997-43-0 | |
| 305.344 | |
| RWEKWRQKAHQYNE-UHFFFAOYSA-N | |
| 7-(bromomethyl)pentadecane |
| C16H33Br | |
| MFCD00136231 | |
| 18459580 | |
| CCCCCCCCC(CCCCCC)CBr |
Specifications
| 52997-43-0 | |
| 118°C | |
| MFCD00136231 | |
| RWEKWRQKAHQYNE-UHFFFAOYSA-N | |
| 7-(bromomethyl)pentadecane | |
| 18459580 | |
| ≥95.0% (GC) | |
| 7-(Bromomethyl)pentadecane |
| Yellow | |
| C16H33Br | |
| 5 mL | |
| CCCCCCCCC(CCCCCC)CBr | |
| 305.344 | |
| 305.34 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : Yes