missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(Bromomethyl)pentadecane 95.0+%, TCI America™
Supplier: TCI America B45445ML
Specifications
| 7-(Bromomethyl)pentadecane | |
| Yellow | |
| C16H33Br | |
| 5 mL | |
| CCCCCCCCC(CCCCCC)CBr | |
| 305.344 | |
| 305.34 | |
| Liquid |
| 52997-43-0 | |
| 118°C | |
| MFCD00136231 | |
| RWEKWRQKAHQYNE-UHFFFAOYSA-N | |
| 7-(bromomethyl)pentadecane | |
| 18459580 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 52997-43-0 | |
| 305.344 | |
| RWEKWRQKAHQYNE-UHFFFAOYSA-N | |
| 7-(bromomethyl)pentadecane |
| C16H33Br | |
| MFCD00136231 | |
| 18459580 | |
| CCCCCCCCC(CCCCCC)CBr |
Safety and Handling
EINECSNumber : (2)-0067
TSCA : Yes