missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3-hydroxy-2-naphthoic Acid 98.0+%, TCI America™

Catalog No. p-7138108
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31801G 1 g
B31805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31801G Supplier TCI America Supplier No. B31801G
Only null left

Chemical Identifiers

CAS 1779-11-9
Molecular Formula C11H7BrO3
Molecular Weight (g/mol) 267.078
MDL Number MFCD00137823
InChI Key XZWXQSGFZHRDNB-UHFFFAOYSA-N
Synonym 7-Bromo-3-hydroxy-2-naphthalenecarboxylic Acid
PubChem CID 74503
IUPAC Name 7-bromo-3-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC(=CC2=CC(=C(C=C21)O)C(=O)O)Br

Specifications

Melting Point 262°C
Color Yellow
Quantity 1 g
Formula Weight 267.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromo-3-hydroxy-2-naphthoic Acid