CAS RN 1779-11-9
CAS RN 1779-11-9
IUPAC Name:
7-bromo-3-hydroxynaphthalene-2-carboxylic acid
Synonyms:
7-bromo-3-hydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol):
267.08
Molecular Formula:
C11H7BrO3
InChi Key:
XZWXQSGFZHRDNB-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(O)C=C2C=CC(Br)=CC2=C1
1
–
1
of
1
results
1
7-Bromo-3-hydroxy-2-naphthoic Acid 98.0+%, TCI America™
CAS: 1779-11-9 Molecular Formula: C11H7BrO3 Molecular Weight (g/mol): 267.078 MDL Number: MFCD00137823 InChI Key: XZWXQSGFZHRDNB-UHFFFAOYSA-N Synonym: 7-Bromo-3-hydroxy-2-naphthalenecarboxylic Acid PubChem CID: 74503 IUPAC Name: 7-bromo-3-hydroxynaphthalene-2-carboxylic acid SMILES: C1=CC(=CC2=CC(=C(C=C21)O)C(=O)O)Br