missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-2-naphthol 97.0+%, TCI America™

Catalog No. p-7123629
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45801G 1 g
B45805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45801G Supplier TCI America Supplier No. B45801G
Only null left

Chemical Identifiers

CAS 116230-30-9
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.069
MDL Number MFCD00277644
InChI Key VWSBGGRCEQOTNU-UHFFFAOYSA-N
PubChem CID 613827
IUPAC Name 7-bromonaphthalen-2-ol
SMILES C1=CC(=CC2=C1C=CC(=C2)Br)O

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 1 g
Formula Weight 223.07
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromo-2-naphthol