accessibility menu, dialog, popup

UserName

BrHO

CAS RN 116230-30-9

IUPAC Name: 7-bromonaphthalen-2-ol
Synonyms: 7-bromonaphthalen-2-ol
Molecular Weight (g/mol): 223.07
Molecular Formula: C10H7BrO
InChi Key: VWSBGGRCEQOTNU-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC(Br)=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results