missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Amino-4-methylcoumarin-3-acetic Acid 95.0+%, TCI America™
Supplier: TCI America A2820250MG
Specifications
| 7-Amino-4-methylcoumarin-3-acetic Acid | |
| C12H11NO4 | |
| 250 mg | |
| QEQDLKUMPUDNPG-UHFFFAOYSA-N | |
| 2-(7-amino-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid | |
| 129367 | |
| ≥95.0% (T) |
| 106562-32-7 | |
| MFCD00081051 | |
| AMCA | |
| CC1=C(CC(O)=O)C(=O)OC2=CC(N)=CC=C12 | |
| 233.22 | |
| 233.22 | |
| Crystalline Powder |
Chemical Identifiers
| 106562-32-7 | |
| 233.22 | |
| QEQDLKUMPUDNPG-UHFFFAOYSA-N | |
| 129367 | |
| CC1=C(CC(O)=O)C(=O)OC2=CC(N)=CC=C12 |
| C12H11NO4 | |
| MFCD00081051 | |
| AMCA | |
| 2-(7-amino-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid |
Safety and Handling
TSCA : No