missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-4-methylcoumarin-3-acetic Acid 95.0+%, TCI America™

Catalog No. p-7118095
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A282050MG 50 mg
A2820250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A282050MG Supplier TCI America Supplier No. A282050MG
Only null left

Chemical Identifiers

CAS 106562-32-7
Molecular Formula C12H11NO4
Molecular Weight (g/mol) 233.22
MDL Number MFCD00081051
InChI Key QEQDLKUMPUDNPG-UHFFFAOYSA-N
Synonym AMCA
PubChem CID 129367
IUPAC Name 2-(7-amino-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid
SMILES CC1=C(CC(O)=O)C(=O)OC2=CC(N)=CC=C12

Specifications

Quantity 50 mg
Formula Weight 233.22
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Amino-4-methylcoumarin-3-acetic Acid