missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine 98.0+%, TCI America™
Supplier: TCI America B4908200MG
Specifications
| 7,7′-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine | |
| 376°C | |
| C36H20N2O4 | |
| HN-D1 | |
| C1=CC=C2C(=C1)N3C4=CC=CC=C4OC5=C3C(=CC(=C5)C6=CC7=C8C(=C6)OC9=CC=CC=C9N8C1=CC=CC=C1O7)O2 | |
| 89981730 | |
| ≥98.0% (HPLC) |
| 1395881-55-6 | |
| Yellow | |
| 200 mg | |
| LQIJYRYGPVWGLO-UHFFFAOYSA-N | |
| 544.566 | |
| 544.57 | |
| Crystalline Powder |
Chemical Identifiers
| 1395881-55-6 | |
| 544.566 | |
| HN-D1 | |
| C1=CC=C2C(=C1)N3C4=CC=CC=C4OC5=C3C(=CC(=C5)C6=CC7=C8C(=C6)OC9=CC=CC=C9N8C1=CC=CC=C1O7)O2 |
| C36H20N2O4 | |
| LQIJYRYGPVWGLO-UHFFFAOYSA-N | |
| 89981730 |
Safety and Handling
TSCA : No