accessibility menu, dialog, popup

UserName

NOONOO

CAS RN 1395881-55-6

IUPAC Name: 11-{8,14-dioxa-1-azapentacyclo[11.7.1.0²,⁷.0⁹,²¹.0¹⁵,²⁰]henicosa-2,4,6,9,11,13(21),15,17,19-nonaen-11-yl}-8,14-dioxa-1-azapentacyclo[11.7.1.0²,⁷.0⁹,²¹.0¹⁵,²⁰]henicosa-2,4,6,9,11,13(21),15,17,19-nonaene
Synonyms: 11-{8,14-dioxa-1-azapentacyclo[11.7.1.0²,⁷.0⁹,²¹.0¹⁵,²⁰]henicosa-2,4,6,9,11,13(21),15,17,19-nonaen-11-yl}-8,14-dioxa-1-azapentacyclo[11.7.1.0²,⁷.0⁹,²¹.0¹⁵,²⁰]henicosa-2,4,6,9,11,13(21),15,17,19-nonaene
Molecular Weight (g/mol): 544.57
Molecular Formula: C36H20N2O4
InChi Key: LQIJYRYGPVWGLO-UHFFFAOYSA-N
SMILES: O1C2=CC=CC=C2N2C3=CC=CC=C3OC3=C2C1=CC(=C3)C1=CC2=C3N(C4=CC=CC=C4OC3=C1)C1=CC=CC=C1O2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results