missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Undecanone 98.0+%, TCI America™
Supplier: TCI America U0043100ML
Specifications
| 6-Undecanone | |
| Yellow | |
| C11H22O | |
| 100 mL | |
| ZPQAKYPOZRXKFA-UHFFFAOYSA-N | |
| undecan-6-one | |
| 13561 | |
| ≥98.0% (GC) |
| 927-49-1 | |
| 226°C | |
| MFCD00009516 | |
| 6-undecanone, dipentyl ketone, amyl ketone, diamyl ketone, pentyl ketone, 6-oxoundecane, 6undecanone, di-n-amyl ketone, ketones, c11, c11 ketones | |
| CCCCCC(=O)CCCCC | |
| 170.296 | |
| 170.30 | |
| Liquid |
Chemical Identifiers
| 927-49-1 | |
| 170.296 | |
| ZPQAKYPOZRXKFA-UHFFFAOYSA-N | |
| 13561 | |
| CCCCCC(=O)CCCCC |
| C11H22O | |
| MFCD00009516 | |
| 6-undecanone, dipentyl ketone, amyl ketone, diamyl ketone, pentyl ketone, 6-oxoundecane, 6undecanone, di-n-amyl ketone, ketones, c11, c11 ketones | |
| undecan-6-one |
Safety and Handling
EINECSNumber : (2)-2475
RTECSNumber : YQ2828000
TSCA : Yes