missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Undecanone 98.0+%, TCI America™
$134.21 - $329.94
Chemical Identifiers
| CAS | 927-49-1 |
|---|---|
| Molecular Formula | C11H22O |
| Molecular Weight (g/mol) | 170.296 |
| MDL Number | MFCD00009516 |
| InChI Key | ZPQAKYPOZRXKFA-UHFFFAOYSA-N |
| Synonym | 6-undecanone, dipentyl ketone, amyl ketone, diamyl ketone, pentyl ketone, 6-oxoundecane, 6undecanone, di-n-amyl ketone, ketones, c11, c11 ketones |
| PubChem CID | 13561 |
| IUPAC Name | undecan-6-one |
| SMILES | CCCCCC(=O)CCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
U004325ML
|
TCI America
U004325ML |
25 mL |
Each for $134.21
|
|
|||||
|
U0043100ML
|
TCI America
U0043100ML |
100 mL |
Each for $329.94
|
|
|||||
Chemical Identifiers
| 927-49-1 | |
| 170.296 | |
| ZPQAKYPOZRXKFA-UHFFFAOYSA-N | |
| 13561 | |
| CCCCCC(=O)CCCCC |
| C11H22O | |
| MFCD00009516 | |
| 6-undecanone, dipentyl ketone, amyl ketone, diamyl ketone, pentyl ketone, 6-oxoundecane, 6undecanone, di-n-amyl ketone, ketones, c11, c11 ketones | |
| undecan-6-one |
Specifications
| 927-49-1 | |
| 226°C | |
| MFCD00009516 | |
| 6-undecanone, dipentyl ketone, amyl ketone, diamyl ketone, pentyl ketone, 6-oxoundecane, 6undecanone, di-n-amyl ketone, ketones, c11, c11 ketones | |
| CCCCCC(=O)CCCCC | |
| 170.296 | |
| 170.30 | |
| Liquid |
| Yellow | |
| C11H22O | |
| 25 mL | |
| ZPQAKYPOZRXKFA-UHFFFAOYSA-N | |
| undecan-6-one | |
| 13561 | |
| ≥98.0% (GC) | |
| 6-Undecanone |
Safety and Handling
EINECSNumber : (2)-2475
RTECSNumber : YQ2828000
TSCA : Yes