missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 6-Thioguanine | |
| Yellow | |
| MFCD00233553 | |
| 2811 | |
| WYWHKKSPHMUBEB-UHFFFAOYSA-N | |
| 2-amino-6,7-dihydro-3H-purine-6-thione | |
| 2723601 | |
| 167.19 | |
| Crystalline Powder |
| 154-42-7 | |
| C5H5N5S | |
| 1 g | |
| 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol | |
| NC1=NC(=S)C2=C(N1)N=CN2 | |
| 167.19 | |
| CHEBI:9555 | |
| ≥95.0% (HPLC) |
Chemical Identifiers
| 154-42-7 | |
| 167.19 | |
| WYWHKKSPHMUBEB-UHFFFAOYSA-N | |
| 2723601 | |
| 2-amino-6,7-dihydro-3H-purine-6-thione |
| C5H5N5S | |
| MFCD00233553 | |
| 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol | |
| CHEBI:9555 | |
| NC1=NC(=S)C2=C(N1)N=CN2 |
Safety and Handling
RTECSNumber : UP0740000
TSCA : No