missing translation for 'onlineSavingsMsg'
Learn More

6-Thioguanine 95.0+%, TCI America™

Catalog No. p-7136148
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T02121G 1 g
T02125G 5 g
2 options

Catalog No. T02121G

Supplier: TCI America T02121G

Only null left

Chemical Identifiers

CAS 154-42-7
Molecular Formula C5H5N5S
Molecular Weight (g/mol) 167.19
MDL Number MFCD00233553
InChI Key WYWHKKSPHMUBEB-UHFFFAOYSA-N
Synonym 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol
PubChem CID 2723601
ChEBI CHEBI:9555
IUPAC Name 2-amino-6,7-dihydro-3H-purine-6-thione
SMILES NC1=NC(=S)C2=C(N1)N=CN2

Specifications

Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 167.19
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Thioguanine
RTECSNumber UP0740000
TSCA No