accessibility menu, dialog, popup

UserName

CAS RN 154-42-7

IUPAC Name: 2-amino-6,7-dihydro-3H-purine-6-thione
Synonyms: thioguanine|6-thioguanine|lanvis|tioguanine|ThG|tabloid, thioguanine|2-amino-6-mercaptopurine|6 mercaptoguanine|tioguanina|thioguanine tabloid
Molecular Weight (g/mol): 167.19
Molecular Formula: C5H5N5S
InChi Key: WYWHKKSPHMUBEB-UHFFFAOYSA-N
SMILES: NC1=NC(=S)C2=C(N1)N=CN2
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)