missing translation for 'onlineSavingsMsg'
Learn More

6-tert-Butyl-2,4-xylenol 97.0+%, TCI America™

Catalog No. B0903500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0903500G 500 g
B090325G 25 g
B0903100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0903500G Supplier TCI America Supplier No. B0903500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1879-09-0 | C12H18O | 178.28 g/mol

Chemical Identifiers

CAS 1879-09-0
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00002234
InChI Key OPLCSTZDXXUYDU-UHFFFAOYSA-N
Synonym 2,4-dimethyl-6-tert-butylphenol, 2-tert-butyl-4,6-dimethylphenol, 6-tert-butyl-2,4-dimethylphenol, topanol a, 6-tert-butyl-2,4-xylenol, 6-t-butyl-2,4-xylenol, prodox 340, phenol, 2-1,1-dimethylethyl-4,6-dimethyl, 6-t-butyl-2,4-dimethylphenol, 2,4-xylenol, 6-tert-butyl
PubChem CID 15884
IUPAC Name 2-tert-butyl-4,6-dimethylphenol
SMILES CC1=CC(C)=C(O)C(=C1)C(C)(C)C

Specifications

Color White-Yellow
Quantity 500 g
Physical Form Crystal-Lump at 20°C
UN Number 2811
Boiling Point 249°C
Formula Weight 178.28
Percent Purity ≥97.0% (GC)
Chemical Name or Material 6-tert-Butyl-2,4-xylenol
EINECSNumber (3)-0540
RTECSNumber ZE6825000
TSCA Yes