accessibility menu, dialog, popup

UserName

CAS RN 1879-09-0

IUPAC Name: 2-tert-butyl-4,6-dimethylphenol
Synonyms: butylated xylenol
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: OPLCSTZDXXUYDU-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(O)C(=C1)C(C)(C)C
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)