missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrophthalide 95.0+%, TCI America™

Catalog No. p-7134102
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10551G 1 g
N10555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10551G Supplier TCI America Supplier No. N10551G
Only null left

Chemical Identifiers

CAS 610-93-5
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00033529
InChI Key RNWGZXAHUPFXLL-UHFFFAOYSA-N
Synonym 6-nitrophthalide, 6-nitroisobenzofuran-1 3h-one, 6-nitro-3h-isobenzofuran-1-one, 6-nitro-1,3-dihydroisobenzofuran-1-one, 6-nitro-1 3h-isobenzofuranone, 6-nitro-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 6-nitro, 6-nitro-3-hydroisobenzofuran-1-one, 6-nitro-1,3-dihydro-2-benzofuran-1-one, 6-nitrophtalide
PubChem CID 223584
IUPAC Name 6-nitro-1,3-dihydro-2-benzofuran-1-one
SMILES [O-][N+](=O)C1=CC=C2COC(=O)C2=C1

Specifications

Melting Point 145°C
Quantity 1 g
Formula Weight 179.13
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitrophthalide