accessibility menu, dialog, popup

UserName

ONOOO

CAS RN 610-93-5

IUPAC Name: 6-nitro-1,3-dihydro-2-benzofuran-1-one
Synonyms: 6-nitro-3H-2-benzofuran-1-one
Molecular Weight (g/mol): 179.13
Molecular Formula: C8H5NO4
InChi Key: RNWGZXAHUPFXLL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2COC(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)