missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrophthalide, 97%

Catalog No. AAB2037514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2037514 25 g
AAB2037506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2037514 Supplier Thermo Scientific Chemicals Supplier No. B2037514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610-93-5 | C8H5NO4 | 179.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610-93-5
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00033529
InChI Key RNWGZXAHUPFXLL-UHFFFAOYSA-N
Synonym 6-nitrophthalide, 6-nitroisobenzofuran-1 3h-one, 6-nitro-3h-isobenzofuran-1-one, 6-nitro-1,3-dihydroisobenzofuran-1-one, 6-nitro-1 3h-isobenzofuranone, 6-nitro-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 6-nitro, 6-nitro-3-hydroisobenzofuran-1-one, 6-nitro-1,3-dihydro-2-benzofuran-1-one, 6-nitrophtalide
PubChem CID 223584
IUPAC Name 6-nitro-3H-2-benzofuran-1-one
SMILES [O-][N+](=O)C1=CC=C2COC(=O)C2=C1

Specifications

Melting Point 140°C to 146°C
Quantity 25 g
Formula Weight 179.13
Percent Purity 97%
Chemical Name or Material 6-Nitrophthalide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 676-656-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only