missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrochromone 98.0+%, TCI America™

$100.67 - $370.95

Chemical Identifiers

CAS 51484-05-0
Molecular Formula C9H5NO4
Molecular Weight (g/mol) 191.142
MDL Number MFCD02954226
InChI Key ORWADBVBOPTYQT-UHFFFAOYSA-N
Synonym 6-nitrochromone, 6-nitro-4h-chromen-4-one, 6-nitrobenzopyran-4-one, acmc-20alxw, 6-nitro-4-oxo-4h-1-benzopyran, 6-nitro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-nitro
PubChem CID 847716
IUPAC Name 6-nitrochromen-4-one
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C=CO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10031G
View Documents
TCI America
N10031G
1 g
Each for $100.67
Only null left
 
N10035G
View Documents
TCI America
N10035G
5 g
Each for $370.95
Only null left
 
Description

Description

Specifications

Chemical Identifiers

51484-05-0
191.142
ORWADBVBOPTYQT-UHFFFAOYSA-N
847716
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C=CO2
C9H5NO4
MFCD02954226
6-nitrochromone, 6-nitro-4h-chromen-4-one, 6-nitrobenzopyran-4-one, acmc-20alxw, 6-nitro-4-oxo-4h-1-benzopyran, 6-nitro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-nitro
6-nitrochromen-4-one

Specifications

51484-05-0
Yellow
MFCD02954226
6-nitrochromone, 6-nitro-4h-chromen-4-one, 6-nitrobenzopyran-4-one, acmc-20alxw, 6-nitro-4-oxo-4h-1-benzopyran, 6-nitro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-nitro
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C=CO2
191.142
191.14
Crystalline Powder
174°C
C9H5NO4
1 g
ORWADBVBOPTYQT-UHFFFAOYSA-N
6-nitrochromen-4-one
847716
≥98.0% (GC)
6-Nitrochromone
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents