accessibility menu, dialog, popup

UserName

NOOOO

CAS RN 51484-05-0

IUPAC Name: 6-nitro-4H-chromen-4-one
Synonyms: 6-nitrochromen-4-one
Molecular Weight (g/mol): 191.14
Molecular Formula: C9H5NO4
InChi Key: ORWADBVBOPTYQT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2OC=CC(=O)C2=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)