missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrochromone 98.0+%, TCI America™

Catalog No. N10035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10035G 5 g
N10031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10035G Supplier TCI America Supplier No. N10035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51484-05-0
Molecular Formula C9H5NO4
Molecular Weight (g/mol) 191.142
MDL Number MFCD02954226
InChI Key ORWADBVBOPTYQT-UHFFFAOYSA-N
Synonym 6-nitrochromone, 6-nitro-4h-chromen-4-one, 6-nitrobenzopyran-4-one, acmc-20alxw, 6-nitro-4-oxo-4h-1-benzopyran, 6-nitro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-nitro
PubChem CID 847716
IUPAC Name 6-nitrochromen-4-one
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C=CO2

Specifications

Melting Point 174°C
Color Yellow
Quantity 5 g
Formula Weight 191.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitrochromone