missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Morpholinonicotinic acid, 95+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC17501DA
Specifications
| 6-Morpholinonicotinic acid | |
| 95% | |
| C10H12N2O3 | |
| 6-morpholinonicotinic acid, 6-morpholin-4-ylnicotinic acid, 6-morpholin-4-yl pyridine-3-carboxylic acid, 6-4-morpholinyl-3-pyridinecarboxylic acid, 6-morpholin-4-yl-nicotinic acid, 3-pyridinecarboxylic acid, 6-4-morpholinyl, 6-morpholinopyridine-3-carboxylic acid, pubchem19467, acmc-209a8e, 3-pyridinecarboxylicacid, 6-4-morpholinyl | |
| C1COCCN1C2=NC=C(C=C2)C(=O)O | |
| 208.217 | |
| 208.22 |
| 120800-52-4 | |
| Amber Glass Bottle | |
| 1g | |
| XXDSDFLDYNISKD-UHFFFAOYSA-N | |
| 6-morpholin-4-ylpyridine-3-carboxylic acid | |
| 2776460 | |
| 95% |
Chemical Identifiers
| 120800-52-4 | |
| 208.217 | |
| 6-morpholinonicotinic acid, 6-morpholin-4-ylnicotinic acid, 6-morpholin-4-yl pyridine-3-carboxylic acid, 6-4-morpholinyl-3-pyridinecarboxylic acid, 6-morpholin-4-yl-nicotinic acid, 3-pyridinecarboxylic acid, 6-4-morpholinyl, 6-morpholinopyridine-3-carboxylic acid, pubchem19467, acmc-209a8e, 3-pyridinecarboxylicacid, 6-4-morpholinyl | |
| 6-morpholin-4-ylpyridine-3-carboxylic acid |
| C10H12N2O3 | |
| XXDSDFLDYNISKD-UHFFFAOYSA-N | |
| 2776460 | |
| C1COCCN1C2=NC=C(C=C2)C(=O)O |