missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Morpholinonicotinic acid, 95+%, Thermo Scientific™
Chemical Identifiers
| CAS | 120800-52-4 |
|---|---|
| Molecular Formula | C10H12N2O3 |
| Molecular Weight (g/mol) | 208.217 |
| InChI Key | XXDSDFLDYNISKD-UHFFFAOYSA-N |
| Synonym | 6-morpholinonicotinic acid, 6-morpholin-4-ylnicotinic acid, 6-morpholin-4-yl pyridine-3-carboxylic acid, 6-4-morpholinyl-3-pyridinecarboxylic acid, 6-morpholin-4-yl-nicotinic acid, 3-pyridinecarboxylic acid, 6-4-morpholinyl, 6-morpholinopyridine-3-carboxylic acid, pubchem19467, acmc-209a8e, 3-pyridinecarboxylicacid, 6-4-morpholinyl |
| PubChem CID | 2776460 |
| IUPAC Name | 6-morpholin-4-ylpyridine-3-carboxylic acid |
| SMILES | C1COCCN1C2=NC=C(C=C2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
CC17501CB
|
Thermo Scientific Chemicals
CC17501CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC17501DA
|
Thermo Scientific Chemicals
CC17501DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC17501DE
|
Thermo Scientific Chemicals
CC17501DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 120800-52-4 | |
| 208.217 | |
| 6-morpholinonicotinic acid, 6-morpholin-4-ylnicotinic acid, 6-morpholin-4-yl pyridine-3-carboxylic acid, 6-4-morpholinyl-3-pyridinecarboxylic acid, 6-morpholin-4-yl-nicotinic acid, 3-pyridinecarboxylic acid, 6-4-morpholinyl, 6-morpholinopyridine-3-carboxylic acid, pubchem19467, acmc-209a8e, 3-pyridinecarboxylicacid, 6-4-morpholinyl | |
| 6-morpholin-4-ylpyridine-3-carboxylic acid |
| C10H12N2O3 | |
| XXDSDFLDYNISKD-UHFFFAOYSA-N | |
| 2776460 | |
| C1COCCN1C2=NC=C(C=C2)C(=O)O |
Specifications
| 120800-52-4 | |
| Amber Glass Bottle | |
| 250mg | |
| XXDSDFLDYNISKD-UHFFFAOYSA-N | |
| 6-morpholin-4-ylpyridine-3-carboxylic acid | |
| 2776460 | |
| 95% |
| 95% | |
| C10H12N2O3 | |
| 6-morpholinonicotinic acid, 6-morpholin-4-ylnicotinic acid, 6-morpholin-4-yl pyridine-3-carboxylic acid, 6-4-morpholinyl-3-pyridinecarboxylic acid, 6-morpholin-4-yl-nicotinic acid, 3-pyridinecarboxylic acid, 6-4-morpholinyl, 6-morpholinopyridine-3-carboxylic acid, pubchem19467, acmc-209a8e, 3-pyridinecarboxylicacid, 6-4-morpholinyl | |
| C1COCCN1C2=NC=C(C=C2)C(=O)O | |
| 208.217 | |
| 208.22 | |
| 6-Morpholinonicotinic acid |