missing translation for 'onlineSavingsMsg'
Learn More

6-(Methylthio)purine, 98%

Catalog No. AC226560010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226560010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC226560010 Supplier Thermo Scientific Chemicals Supplier No. 226560010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-66-8 | C6H6N4S | 166.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-66-8
Molecular Formula C6H6N4S
Molecular Weight (g/mol) 166.20
MDL Number MFCD00005576
InChI Key UIJIQXGRFSPYQW-UHFFFAOYSA-N
Synonym 6-methylthio purine, 6-methylmercaptopurine, 6-methylthiopurine, 1h-purine, 6-methylthio, 6-methyl mp, purine, 6-methylthio, s-methyl-6-mercaptopurine, 6-methylsulfanyl-9h-purine, unii-6v404dv25o, thiopurine s-methylether
PubChem CID 5778
ChEBI CHEBI:28279
IUPAC Name 6-(methylsulfanyl)-7H-purine
SMILES CSC1=C2NC=NC2=NC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 219.0°C to 223.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 166.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-(Methylthio)purine, 98%
Show More Show Less
EINECSNumber 200-057-3

RUO – Research Use Only