accessibility menu, dialog, popup

UserName

CAS RN 50-66-8

IUPAC Name: 6-(methylsulfanyl)-7H-purine
Synonyms: 6-(methylthio)purine
Molecular Weight (g/mol): 166.2
Molecular Formula: C6H6N4S
InChi Key: UIJIQXGRFSPYQW-UHFFFAOYSA-N
SMILES: CSC1=C2NC=NC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results