missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxy-1-tetralone 99.0+%, TCI America™
Supplier: TCI America M0787250G
Specifications
| 6-Methoxy-1-tetralone | |
| 79°C | |
| MFCD00001695 | |
| 6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone | |
| COC1=CC2=C(C=C1)C(=O)CCC2 | |
| 176.215 | |
| 176.22 | |
| Crystalline Powder |
| 1078-19-9 | |
| C11H12O2 | |
| 250 g | |
| MNALUTYMBUBKNX-UHFFFAOYSA-N | |
| 6-methoxy-3,4-dihydro-2H-naphthalen-1-one | |
| 14112 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 1078-19-9 | |
| 176.215 | |
| MNALUTYMBUBKNX-UHFFFAOYSA-N | |
| 14112 | |
| COC1=CC2=C(C=C1)C(=O)CCC2 |
| C11H12O2 | |
| MFCD00001695 | |
| 6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone | |
| 6-methoxy-3,4-dihydro-2H-naphthalen-1-one |
Safety and Handling
EINECSNumber : (4)-0371
TSCA : Yes