missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1-tetralone 99.0+%, TCI America™

Catalog No. p-7132361
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
M078725G 25 g
M0787250G 250 g
2 options

Catalog No. M078725G

Supplier: TCI America M078725G

Only null left

Chemical Identifiers

CAS 1078-19-9
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00001695
InChI Key MNALUTYMBUBKNX-UHFFFAOYSA-N
Synonym 6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone
PubChem CID 14112
IUPAC Name 6-methoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(C=C1)C(=O)CCC2

Specifications

Melting Point 79°C
Quantity 25 g
Formula Weight 176.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-1-tetralone
EINECSNumber (4)-0371
TSCA Yes