missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1,2,3,4-tetrahydronaphthalene 98.0+%, TCI America™

Catalog No. M07655ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07655ML 5 mL
M076525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07655ML Supplier TCI America Supplier No. M07655ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1730-48-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00001737
InChI Key SUFDFKRJYKNTFH-UHFFFAOYSA-N
Synonym 6-methoxytetralin, naphthalene, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxynaphthalene, unii-d0xg2umz29, d0xg2umz29, methyl 1,2,3,4-tetrahydro-6-naphthyl ether, acmc-20aj41, 1,3,4-tetrahydro-6-methoxynaphthalene, 6-methoxy-1,3,4-tetrahydronaphthalene, 7-methoxy-3,4-dihydro-1h-naphthalene
PubChem CID 74414
IUPAC Name 6-methoxy-1,2,3,4-tetrahydronaphthalene
SMILES COC1=CC=C2CCCCC2=C1

Specifications

Color Yellow
Boiling Point 131°C
Quantity 5 mL
Formula Weight 162.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Methoxy-1,2,3,4-tetrahydronaphthalene
EINECSNumber (4)-1394
TSCA Yes