accessibility menu, dialog, popup

UserName

CAS RN 1730-48-9

IUPAC Name: 6-methoxy-1,2,3,4-tetrahydronaphthalene
Synonyms: 2-methoxy-5,6,7,8-tetrahydronaphthalene
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: SUFDFKRJYKNTFH-UHFFFAOYSA-N
SMILES: COC1=CC=C2CCCCC2=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results