missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxy-2-naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™
$106.25 - $106.25
Specifications
| CAS | 173194-95-1 |
|---|---|
| Melting Point | 224°C |
| Molecular Formula | C10H9BO3 |
| MDL Number | MFCD00092986 |
| InChI Key | SNJANQMHRGSHFN-UHFFFAOYSA-N |
Specifications
| 173194-95-1 | |
| C10H9BO3 | |
| SNJANQMHRGSHFN-UHFFFAOYSA-N | |
| (6-hydroxynaphthalen-2-yl)boronic acid | |
| 4349738 | |
| Crystalline Powder |
| 224°C | |
| MFCD00092986 | |
| B(C1=CC2=C(C=C1)C=C(C=C2)O)(O)O | |
| 187.989 | |
| 187.99 |