accessibility menu, dialog, popup

UserName

CAS RN 173194-95-1

BOHOHHO

CAS RN 173194-95-1

IUPAC Name: (6-hydroxynaphthalen-2-yl)boronic acid
Synonyms: 6-hydroxynaphthalen-2-ylboronic acid
Molecular Weight (g/mol): 187.99
Molecular Formula: C10H9BO3
InChi Key: SNJANQMHRGSHFN-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2C=C(O)C=CC2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results