missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ 6 ft. Logic™ Class II, Type A2 Biosafety Cabinet, US Model
Logic Class II, Type A2 Biosafety Cabinets offer personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. All models ensure the highest standard of safety and are listed to NSF/ANSI 49. Logic cabinets maximize safety, performance, and comfort.
$31,637.85 - $34,801.65
Specifications
| Description | 6 ft. Logic Class II, Type A2 Biosafety Cabinet |
|---|---|
| Chamber Material | Stainless Steel (304) |
| Width (Metric) Interior | 184.2 cm |
| Product Type | Class II, Type A2 Bio Safety Cabinet |
| Model | Logic A2 |
| Catalog Number | Mfr. No. | Includes | Plug Type | Sash Opening (Metric) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Plug Type | Sash Opening (Metric) | Price | Quantity & Availability | |||
|
01812573
|
Labconco™
302680002 |
Cabinet | North America (115 V, 20 A) | 20.3 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01812571
|
Labconco™
302680102 |
Cabinet, Base Stand | North America (115 V, 20 A) | 20.3 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01812569
|
Labconco™
302681002 |
Cabinet, Accessory Package | North America (115 V, 20 A) | 20.3 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01812567
|
Labconco™
302681102 |
Cabinet, Accessory Package, Base Stand | North America (115 V, 20 A) | 20.3 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01812589
|
Labconco™
302610002 |
Cabinet | North America (115 V, 20 A) | 25.4 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01812587
|
Labconco™
302610102 |
Cabinet, Base Stand | North America (115 V, 20 A) | 25.4 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01812585
|
Labconco™
302611002 |
Cabinet, Accessory Package | North America (115 V, 20 A) | 25.4 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01812583
|
Labconco™
302611102 |
Cabinet, Accessory Package, Base Stand | North America (115 V, 20 A) | 25.4 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01812577
|
Labconco™
302621002 |
Cabinet, Accessory Package | North America (115 V, 20 A) | 30.5 cm |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Performance Features:
- Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
- Logic OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
- Full, six-year warranty on all Class II biosafety cabinets
- Optional Configurations
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Telescoping base stand with shelf and leveling feet
Specifications
| 6 ft. Logic Class II, Type A2 Biosafety Cabinet | |
| 184.2 cm | |
| Logic A2 | |
| 2 Double, Left and Right Side | |
| 78.4 in. | |
| 99.99% | |
| 199.1 cm | |
| 157.2 cm | |
| 72.5 in. | |
| 22.5 in. | |
| 65.3 cm | |
| 69 cm | |
| 81.3 cm |
| Stainless Steel (304) | |
| Class II, Type A2 Bio Safety Cabinet | |
| 2 Duplex | |
| 61.9 in. | |
| 5 in. touchscreen | |
| HEPA | |
| 66 W | |
| 6 Years | |
| 25.7 in. | |
| 27 in. | |
| 57.2 cm | |
| 32 in. |