missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ 6 ft. Logic™ Class II, Type A2 Biosafety Cabinet, US Model
Logic Class II, Type A2 Biosafety Cabinets offer personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. All models ensure the highest standard of safety and are listed to NSF/ANSI 49. Logic cabinets maximize safety, performance, and comfort.
Supplier: Labconco™ 302621002
Description
Performance Features:
- Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
- Logic OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
- Full, six-year warranty on all Class II biosafety cabinets
- Optional Configurations
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Telescoping base stand with shelf and leveling feet
Specifications
| 6 ft. Logic Class II, Type A2 Biosafety Cabinet | |
| Stainless Steel (304) | |
| Class II, Type A2 Bio Safety Cabinet | |
| 67 dBA | |
| North America (115 V, 20 A) | |
| 2 Double, Left and Right Side | |
| 30.5 cm | |
| 1551 m³/hr. | |
| 99.99% | |
| 199.1 cm | |
| 66 W | |
| 157.2 cm | |
| 1 | |
| 6 Years | |
| 60 Hz | |
| 22.5 in. | |
| 65.3 cm | |
| 69 cm | |
| 81.3 cm | |
| 490 W |
| NSF/ANSI 49, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
| 184.2 cm | |
| Logic A2 | |
| 2 Duplex | |
| 490 W | |
| 61.9 in. | |
| 78.4 in. | |
| 5 in. touchscreen | |
| HEPA | |
| Cabinet, Accessory Package | |
| LED, UV | |
| 12 in. | |
| 913 CFM | |
| 72.5 in. | |
| 25.7 in. | |
| 27 in. | |
| 57.2 cm | |
| 32 in. | |
| 115 V |