missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-m-anisaldehyde 95.0+%, TCI America™

Catalog No. F03835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03835G 5 g
F038325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03835G Supplier TCI America Supplier No. F03835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105728-90-3
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00070795
InChI Key DKIQXHIAEMGZGO-UHFFFAOYSA-N
Synonym 6-fluoro-m-anisaldehyde, 2-fluoro-5-methoxy-benzaldehyde, 2-fluoro-5-methoxy benzaldehyde, benzaldehyde, 2-fluoro-5-methoxy, pubchem2634, 4-fluoro-3-formylanisole, acmc-2098hi, ksc490m2b, 2-fluoro-5-methyloxy benzaldehyde, 2-fluoro-5-methoxybenzaldehyde
PubChem CID 2734872
IUPAC Name 2-fluoro-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)F)C=O

Specifications

Color White-Yellow
Boiling Point 230°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Fluoro-m-anisaldehyde